Assay Details
| Gene Symbol: Dcaf12l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190718.1 | NP_001177647.1 | 2 | 2980 | 105 | |
| NM_178739.6 | NP_848854.3 | 2 | 2994 | 105 | ||
| GenBank mRNA | AK034343.1 | - | 2 | 2834 | 105 | |
| AK038557.1 | - | 2 | 2949 | 105 | ||
| BC068319.1 | - | 2 | 2798 | 30431243 | 105 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32351251 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32351253 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32351249 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32351247 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32351250 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020497 | BC068319.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020497 | BB337276.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020497 | AK038557.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020497 | NM_178739.6 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020497 | NM_001190718.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364316 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204985 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140505 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020497 | BC068319.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020497 | BB337276.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020497 | AK038557.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020497 | NM_178739.6 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020497 | NM_001190718.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32351278 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140505 | - | - |
| Whole Mouse Genome G4122A | A_51_P170795 | BC068319.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170795 | BB337276.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170795 | AK038557.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170795 | NM_178739.6 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170795 | NM_001190718.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102819 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140505 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15162870 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15162869 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15162894 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14685443 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14685431 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15162882 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15162883 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32351249 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32351253 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32351250 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32351251 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32351247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32351226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351271 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15162905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14685436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14685424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14685433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14959485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15162875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14629584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15162889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32351200 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10604335 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604336 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604334 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454869_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434170_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10604333 | - | - |
| Mouse Genome 430 2.0 Array | 1454869_at | - | - |
| Mouse Genome 430 2.0 Array | 1454869_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434170_at | - | - |
| Mouse Genome 430 2.0 Array | 1434170_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245404_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245404_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00013545.mm.1 | NM_178739.6.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013545.mm.1 | NM_178739.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013545.mm.1 | TC0X00002136.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00013541.mm.1 | NM_001190718.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013541.mm.1 | NM_001190718.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013541.mm.1 | NM_178739.6.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013541.mm.1 | NM_178739.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013541.mm.1 | TC0X00002136.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00013543.mm.1 | NM_001190718.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013543.mm.1 | NM_001190718.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013543.mm.1 | NM_178739.6.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013543.mm.1 | NM_178739.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013543.mm.1 | TC0X00002136.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00013544.mm.1 | NM_001190718.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013544.mm.1 | NM_001190718.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013544.mm.1 | NM_178739.6.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013544.mm.1 | NM_178739.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00013544.mm.1 | TC0X00002136.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434170_at | - | - |
| Mouse Expression Set 430 | 1434170_PM_at | - | - |
| Mouse Expression Set 430 | 1454869_at | - | - |
| Mouse Expression Set 430 | 1454869_PM_at | - | - |
| Murine Genome U74v2 Set | 170420_PM_r_at | - | - |
| Murine Genome U74v2 Set | 170013_PM_i_at | - | - |
| Murine Genome U74v2 Set | 170013_i_at | - | - |
| Murine Genome U74v2 Set | 170420_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454869_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434170_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454869_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434170_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 488214 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3780288 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003780288 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2497615 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2497615 | - | - |
| mouse-6 v1.1 (Illumina) | scl54299.2.1_23-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3780288 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl54299.2.1_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3780288 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2497615 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2497615 | - | - |

Dcaf12l1
Genomic Map