Assay Details
| Gene Symbol: Gpr135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181752.1 | NP_861417.1 | 1 | 1146 | 79 | |
| GenBank mRNA | AK132237.1 | - | 1282 | 79 | ||
| AK141053.1 | - | 1 | 1230 | 79 | ||
| AY288425.1 | - | 1 | 1146 | 79 | ||
| BC062104.1 | - | 1179 | 6809932 | 79 | ||
| BC114990.1 | - | 1 | 1145 | 79 | ||
| BC115824.1 | - | 1 | 1145 | 79 | ||
| BC139332.1 | - | 1 | 1208 | 8862354 | 79 | |
| BC139335.1 | - | 1 | 1208 | 8862357 | 79 | |
| BC148305.2 | - | 1 | 1146 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530002 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28529998 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530000 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28529996 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28529995 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28529999 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28529997 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530004 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530001 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530003 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275678 | BC139332.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275678 | BC139335.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275678 | BC148305.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275678 | BC062104.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275678 | BC114990.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275678 | BC139332.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275678 | BC139335.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275678 | BC148305.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275678 | BC062104.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P275678 | BC114990.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 671818 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 328365 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 319360 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148608 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275678 | BC139332.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275678 | BC139335.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275678 | BC148305.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275678 | BC062104.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275678 | BC114990.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275678 | BC139332.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275678 | BC139335.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275678 | BC148305.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275678 | BC062104.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P275678 | BC114990.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275678 | BC139332.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275678 | BC139335.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275678 | BC148305.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275678 | BC062104.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275678 | BC114990.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275678 | BC139332.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275678 | BC139335.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275678 | BC148305.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275678 | BC062104.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P275678 | BC114990.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28529996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28529989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28529995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28529998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28529987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28529986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28529993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28529988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28529985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28529984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28529991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28529997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28530005 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148608 | - | - |
| Whole Mouse Genome G4122A | A_52_P275678 | BC139332.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275678 | BC139335.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275678 | BC148305.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275678 | BC062104.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275678 | BC114990.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275678 | BC139332.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275678 | BC139335.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275678 | BC148305.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275678 | BC062104.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P275678 | BC114990.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118250 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148608 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09772240 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09983575 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10372971 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09772238 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28529999 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28529996 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28529998 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530001 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530003 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28529995 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28529997 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530004 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530000 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28530002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28530067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28529963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09772240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10181839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10305201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10305210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10120996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09772238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10181838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10372975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28529977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09983575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28530011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10305200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10112864 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10400895 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439346_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459274_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441944_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10400894 | - | - |
| Mouse Genome 430 2.0 Array | 1441944_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1439346_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459274_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441944_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1459274_at | - | - |
| Mouse Genome 430 2.0 Array | 1439346_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200013020.mm.1 | TC1200001898.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200013021.mm.1 | TC1200001898.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200013019.mm.1 | NM_181752.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013019.mm.1 | NM_181752.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013019.mm.1 | TC1200001898.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439346_PM_at | - | - |
| Mouse Expression Set 430 | 1459274_at | - | - |
| Mouse Expression Set 430 | 1441944_PM_s_at | - | - |
| Mouse Expression Set 430 | 1459274_PM_at | - | - |
| Mouse Expression Set 430 | 1441944_s_at | - | - |
| Mouse Expression Set 430 | 1439346_at | - | - |
| Murine Genome U74v2 Set | 166341_r_at | - | - |
| Murine Genome U74v2 Set | 166341_PM_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441944_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439346_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459274_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441944_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439346_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459274_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 350024 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5130112 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5130112 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005130112 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2659724 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2659724 | - | - |
| mouse-6 v1.1 (Illumina) | scl00238252.1_311-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5130112 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00238252.1_311-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5130112 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2659724 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2659724 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00238252.1_311-S | - | - |

Gpr135
Genomic Map