Assay Details
| Gene Symbol: Klk1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010645.2 | NP_034775.1 | 2 - 3 | 221 | 117 | |
| GenBank mRNA | BC026378.1 | - | 2 - 3 | 221 | 4165617 | 117 |
More Information
Updated Mapping Information
Set Membership: |
|

Klk1b1
Genomic Map