| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900006178.mm.1 |
NM_001162917.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006178.mm.1 |
NM_001162917.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006178.mm.1 |
TC0900000824.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006177.mm.1 |
NM_001162917.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006177.mm.1 |
NM_001162917.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006177.mm.1 |
TC0900000824.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006175.mm.1 |
NM_001162917.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006175.mm.1 |
NM_001162917.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006175.mm.1 |
TC0900000824.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006176.mm.1 |
NM_001162917.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006176.mm.1 |
NM_001162917.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006176.mm.1 |
TC0900000824.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006174.mm.1 |
NM_001162917.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006174.mm.1 |
NM_001162917.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006174.mm.1 |
TC0900000824.mm.1 |
MTA TC |