Assay Details
| Gene Symbol: Prox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175198.4 | NP_780407.1 | 3 - 4 | 1750 | 90 | |
| GenBank mRNA | AK029978.1 | - | 3 - 4 | 1707 | 90 | |
| AK051700.1 | - | 3 - 4 | 1751 | 90 | ||
| BC119330.1 | - | 2 - 3 | 1494 | 8734079 | 90 | |
| BC119332.1 | - | 2 - 3 | 1494 | 8734081 | 90 | |
More Information
Set Membership: |
|

Prox2
Genomic Map