Assay Details
| Gene Symbol: Prokr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144944.3 | NP_659193.3 | 1 - 2 | 197 | 73 | |
| GenBank mRNA | AK041586.1 | - | 1 - 2 | 121 | 73 | |
| AK082195.1 | - | 180 | 73 | |||
| BC043116.1 | - | 1 - 2 | 150 | 6408655 | 73 | |
| BC057557.1 | - | 1 - 2 | 150 | 6408655 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041586.1, BC043116.1, BC057557.1, NM_144944.3 transcripts.
Set Membership: |
|

Prokr2
Genomic Map