Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300010524.mm.1 |
NM_023332.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010524.mm.1 |
TC1300001692.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300010526.mm.1 |
NM_023332.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010526.mm.1 |
TC1300001692.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300010527.mm.1 |
NM_023332.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010527.mm.1 |
TC1300001692.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300010523.mm.1 |
NM_023332.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010523.mm.1 |
TC1300001692.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300010525.mm.1 |
NM_023332.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010525.mm.1 |
TC1300001692.mm.1 |
MTA TC |