Assay Details
| Gene Symbol: Cdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007659.3 | NP_031685.2 | 1 - 2 | 82 | 121 | |
| GenBank mRNA | AK030231.1 | - | 1 - 2 | 83 | 121 | |
| AK051859.1 | - | 1 - 2 | 109 | 121 | ||
| AK135516.1 | - | 1 - 2 | 86 | 121 | ||
| AK168054.1 | - | 1 - 2 | 83 | 121 | ||
| BC024396.1 | - | 1 - 2 | 60 | 4977366 | 121 | |
| U58633.1 | - | 1 - 2 | 86 | 121 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030231.1, AK135516.1, AK168054.1, BC024396.1, NM_007659.3, U58633.1 transcripts.
Set Membership: |
|

Cdk1
Genomic Map