Assay Details
| Gene Symbol: Cdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007659.3 | NP_031685.2 | 5 - 6 | 595 | 76 | |
| GenBank mRNA | AK030231.1 | - | 5 - 6 | 596 | 76 | |
| AK135516.1 | - | 5 - 6 | 599 | 76 | ||
| AK168054.1 | - | 5 - 6 | 596 | 76 | ||
| BC005614.1 | - | 4 - 5 | 488 | 76 | ||
| BC024396.1 | - | 5 - 6 | 573 | 4977366 | 76 | |
| M38724.1 | - | 4 - 5 | 516 | 76 | ||
| U58633.1 | - | 5 - 6 | 599 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk1
Genomic Map