Assay Details
Gene Symbol: Fam89b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023166.2 | NP_075655.2 | 1 - 2 | 432 | 73 | |
GenBank mRNA | AF212921.1 | - | 1 - 2 | 399 | 73 | |
AK003883.1 | - | 1 - 2 | 421 | 73 | ||
BC056166.1 | - | 1 - 2 | 417 | 5715229 | 73 | |
CJ132526.1 | - | 1 - 2 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31930486 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31930485 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31930483 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31930487 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31930484 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2064876 | BC056166.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2064876 | BC013894.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2064876 | BC030163.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2064876 | AK003883.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2064876 | AF212920.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135351 | AF212920.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135351 | AF212921.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 147046 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781704 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 117783 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135351 | AF212920.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135351 | AF212921.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2064876 | BC056166.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2064876 | BC013894.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2064876 | BC030163.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2064876 | AK003883.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2064876 | AF212920.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31930480 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31930472 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31930490 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31930489 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31930491 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31930473 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31930478 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31930483 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31930484 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31930486 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31930475 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31930481 | - | - |
Whole Mouse Genome G4122A | A_51_P464992 | BC056166.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464992 | BC013894.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464992 | BC030163.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464992 | AK003883.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P464992 | AF212920.1 | mouse_transcriptome |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14251866 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14251870 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14251898 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14251911 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14251879 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31930483 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31930487 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31930484 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31930485 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31930486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930489 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31930466 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31930481 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14251883 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31930491 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31930475 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31930486 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31930489 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31930473 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31930478 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14251897 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31930484 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10465216 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465217 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465211 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465213 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465214 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465212 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465210 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416914_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10465215 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10465209 | - | - |
Mouse Genome 430 2.0 Array | 1416914_s_at | - | - |
Mouse Genome 430 2.0 Array | 1416914_PM_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17826_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17826_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1900008840.mm.1 | NM_023166.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900008840.mm.1 | NM_023166.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900008840.mm.1 | NM_181452.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900008840.mm.1 | NM_181452.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900008840.mm.1 | TC1900000981.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1900008837.mm.1 | NM_023166.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900008837.mm.1 | NM_023166.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900008837.mm.1 | NM_181452.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900008837.mm.1 | NM_181452.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900008837.mm.1 | TC1900000981.mm.1 | MTA TC |
Mouse Expression Set 430 | 1416914_s_at | - | - |
Mouse Expression Set 430 | 1416914_PM_s_at | - | - |
Murine Genome U74v2 Set | 96718_at | - | - |
Murine Genome U74v2 Set | 96718_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1416914_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1416914_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416914_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416914_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 916704 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6760408 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6760408 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006760408 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229404 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2680866 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1229404 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2680866 | - | - |
mouse-6 v1.1 (Illumina) | scl017826.2_113-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6760408 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl017826.2_113-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6760408 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3108203 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2680872 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2680872 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3108203 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl017826.2_113-S | - | - |
