Assay Details
Gene Symbol: Uqcrq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025352.2 | NP_079628.1 | 1 - 2 | 300 | 63 | |
GenBank mRNA | AK002389.1 | - | 2 - 3 | 196 | 63 | |
AK003160.1 | - | 2 - 3 | 212 | 63 | ||
AK005374.1 | - | 282 | 63 | |||
AK007982.1 | - | 1 - 2 | 261 | 63 | ||
AK203034.1 | - | 1 - 2 | 48 | 63 | ||
CB950804.1 | - | 1 - 2 | 303 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804725 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804721 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804727 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804720 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804722 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804724 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804726 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804719 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804723 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098802 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965378 | NM_025352.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965378 | AA510422.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965378 | AK005374.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 202165 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165066 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098802 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965378 | NM_025352.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965378 | AA510422.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965378 | AK005374.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P370484 | AK203034.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P370484 | BC027809.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P370484 | BC028519.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P370484 | NM_025352.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P370484 | AA510422.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804733 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804726 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804738 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804721 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804716 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804712 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804708 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804706 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804729 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804731 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804710 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804705 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804743 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804719 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804744 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804709 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804723 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804725 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165066 | - | - |
Whole Mouse Genome G4122A | A_52_P370484 | AK203034.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P370484 | BC027809.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P370484 | BC028519.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P370484 | NM_025352.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P370484 | AA510422.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165066 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09558871 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08997615 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08997613 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08997605 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09558868 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09558870 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08997610 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09379777 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08997601 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804726 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804724 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804725 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804720 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804723 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804719 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804722 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804727 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27804721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804780 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27804716 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27804725 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27804729 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27804714 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27804721 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27804726 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08997615 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08997610 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09190829 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27804733 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27804717 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27804712 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10385820 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10385821 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452613_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1430326_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427880_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10385818 | - | - |
Mouse Genome 430 2.0 Array | 1427880_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1430326_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1437134_at | - | - |
Mouse Genome 430 2.0 Array | 1427880_at | - | - |
Mouse Genome 430 2.0 Array | 1430326_s_at | - | - |
Mouse Genome 430 2.0 Array | 1452613_at | - | - |
Mouse Genome 430 2.0 Array | 1452613_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1437134_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22272_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22272_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100024807.mm.1 | NM_025352.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100024807.mm.1 | TC1100002729.mm.1 | MTA TC |
Mouse Expression Set 430 | 1452613_PM_at | - | - |
Mouse Expression Set 430 | 1452613_at | - | - |
Mouse Expression Set 430 | 1427880_at | - | - |
Mouse Expression Set 430 | 1427880_PM_at | - | - |
Murine Genome U74v2 Set | 93844_at | - | - |
Murine Genome U74v2 Set | 93844_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1427880_at | - | - |
Mouse Genome 430A 2.0 Array | 1427880_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1452613_at | - | - |
Mouse Genome 430A 2.0 Array | 1452613_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1430326_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452613_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427880_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1430326_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427880_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452613_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 780025 | AK038772.1 | genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip | 780025 | AK086391.1 | genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip | 780025 | NM_175280.3 | refseq_mrna_mouse_NM |
Illumina mouseRef-8 v1.0 expression beadchip | 0000780025 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2714442 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2714442 | - | - |
mouse-6 v1.1 (Illumina) | scl40237.1_16-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 780025 | AK038772.1 | genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array | 780025 | AK086391.1 | genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array | 780025 | NM_175280.3 | refseq_mrna_mouse_NM |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl40237.1_16-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 780025 | AK038772.1 | genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 780025 | AK086391.1 | genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 780025 | NM_175280.3 | refseq_mrna_mouse_NM |
MouseRef-8 v2 Expression BeadChip | ILMN_2714442 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2714442 | - | - |
