Assay Details
| Gene Symbol: Mrpl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026310.3 | NP_080586.1 | 1 - 2 | 144 | 116 | |
| GenBank mRNA | AK003104.1 | - | 1 - 2 | 149 | 116 | |
| AK007465.1 | - | 1 - 2 | 157 | 116 | ||
| AK008680.1 | - | 1 - 2 | 145 | 116 | ||
| AK010150.1 | - | 1 - 2 | 197 | 116 | ||
| BC011425.1 | - | 1 - 2 | 119 | 3603233 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055437 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055441 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055444 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055434 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055439 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055435 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055438 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055442 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055436 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P210474 | NM_026310.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P210474 | AK003104.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P210474 | AK008680.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P210474 | AK089039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P210474 | NM_026310.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P210474 | AK003104.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P210474 | AK008680.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P210474 | AK089039.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 466301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208385 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 276711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 27899 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P210474 | NM_026310.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P210474 | AK003104.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P210474 | AK008680.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P210474 | AK089039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P210474 | NM_026310.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P210474 | AK003104.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P210474 | AK008680.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P210474 | AK089039.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P210474 | NM_026310.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P210474 | AK003104.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P210474 | AK008680.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P210474 | AK089039.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P210474 | NM_026310.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P210474 | AK003104.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P210474 | AK008680.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P210474 | AK089039.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31055453 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03282932 | - | - |
| Whole Mouse Genome G4122A | A_51_P210474 | NM_026310.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P210474 | AK003104.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P210474 | AK008680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P210474 | AK089039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P210474 | NM_026310.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P210474 | AK003104.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P210474 | AK008680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P210474 | AK089039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P649258 | AK007465.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P495243 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127232 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13457682 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13457700 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13457696 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13457690 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055436 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055438 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055441 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055439 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055434 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055437 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055442 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055444 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31055435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31055516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055455 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13226709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13129787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13069353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13558330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13336850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055473 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13415393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13415838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13457696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13336851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31055460 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10447883 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447881 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447882 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10473251 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447884 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448373_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10447880 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10473250 | - | - |
| Mouse Genome 430 2.0 Array | 1448373_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448373_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67681_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67681_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700013013.mm.1 | NM_026310.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013013.mm.1 | NM_026310.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013013.mm.1 | TC1700001457.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700013021.mm.1 | NM_026310.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013021.mm.1 | NM_026310.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013021.mm.1 | TC1700001457.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700013012.mm.1 | NM_026310.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013012.mm.1 | NM_026310.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013012.mm.1 | TC1700001457.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700013014.mm.1 | NM_026310.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013014.mm.1 | NM_026310.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013014.mm.1 | TC1700001457.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700013011.mm.1 | NM_026310.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013011.mm.1 | NM_026310.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013011.mm.1 | TC1700001457.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700013010.mm.1 | NM_026310.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013010.mm.1 | NM_026310.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013010.mm.1 | TC1700001457.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700013019.mm.1 | NM_026310.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013019.mm.1 | NM_026310.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700013019.mm.1 | TC1700001457.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448373_at | - | - |
| Mouse Expression Set 430 | 1448373_PM_at | - | - |
| Murine Genome U74v2 Set | 93786_PM_i_at | - | - |
| Murine Genome U74v2 Set | 93786_i_at | - | - |
| Murine Genome U74v2 Set | 93787_f_at | - | - |
| Murine Genome U74v2 Set | 93787_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448373_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448373_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448373_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448373_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 686285 | - | - |
| ABI Mouse Genome Survey Microarray | 864277 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 770433 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000770433 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2659762 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2659762 | - | - |
| mouse-6 v1.1 (Illumina) | scl067681.3_8-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 770433 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067681.3_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 770433 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2823250 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2659762 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2823250 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2659762 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067681.3_8-S | - | - |

Mrpl18
Genomic Map