Assay Details
Gene Symbol: Smim12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030252.2 | NP_084528.1 | 2 | 806 | 89 | |
GenBank mRNA | AK131844.1 | - | 1 | 284 | 89 | |
AK131915.1 | - | 2 | 771 | 89 | ||
AK141221.1 | - | 2 | 740 | 89 | ||
BC147067.1 | - | 2 | 768 | 9056070 | 89 | |
BC147068.1 | - | 2 | 768 | 9056074 | 89 | |
BQ173894.1 | - | 1 | 89 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284268 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284267 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284269 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284266 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187171 | AK141221.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187171 | BC147067.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187171 | BC147068.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187171 | BQ173894.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187171 | NM_030252.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187171 | AK141221.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187171 | BC147067.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187171 | BC147068.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187171 | BQ173894.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187171 | NM_030252.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 433658 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137904 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105203 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187171 | AK141221.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187171 | BC147067.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187171 | BC147068.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187171 | BQ173894.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187171 | NM_030252.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187171 | AK141221.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187171 | BC147067.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187171 | BC147068.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187171 | BQ173894.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187171 | NM_030252.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284279 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284247 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284254 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284266 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284244 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284248 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284270 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284263 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284272 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284258 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284276 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284273 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284284 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284268 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284281 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284264 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284241 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284271 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284274 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284243 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284245 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23284256 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105203 | - | - |
Whole Mouse Genome G4122A | A_51_P187171 | AK141221.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187171 | BC147067.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187171 | BC147068.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187171 | BQ173894.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187171 | NM_030252.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187171 | AK141221.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187171 | BC147067.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187171 | BC147068.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187171 | BQ173894.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187171 | NM_030252.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102185 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105203 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03753045 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03854443 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03656120 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03753044 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03753046 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03854451 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284269 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284268 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284266 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23284267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23284244 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03856824 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284339 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284281 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284325 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284318 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03753043 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03854450 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284298 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284265 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03854451 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284305 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03370800 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03508504 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284343 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284232 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03224724 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284320 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03998355 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284235 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03753044 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03508507 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284341 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284316 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03854445 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03340750 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03998370 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284286 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03720527 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03998357 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03217620 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284241 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284245 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03217621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03753049 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284285 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284314 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03351621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284304 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04007025 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03516659 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03711093 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284342 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284335 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03224725 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03854443 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03998368 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284276 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284264 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03854447 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04022282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284309 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03508502 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03508511 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284306 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03426114 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284344 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284324 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284283 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23284256 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10508271 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10508270 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449189_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10508269 | - | - |
Mouse Genome 430 2.0 Array | 1449189_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1449189_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 80284_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 80284_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400011382.mm.1 | NM_030252.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011382.mm.1 | NM_030252.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011382.mm.1 | TC0400001487.mm.1 | MTA TC |
Mouse Expression Set 430 | 1449189_PM_at | - | - |
Mouse Expression Set 430 | 1449189_at | - | - |
Mouse Genome 430A 2.0 Array | 1449189_at | - | - |
Mouse Genome 430A 2.0 Array | 1449189_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449189_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449189_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 384014 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6900059 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006900059 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1247245 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1247245 | - | - |
mouse-6 v1.1 (Illumina) | scl24944.2_480-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6900059 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl24944.2_480-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900059 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1247245 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1247245 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl24944.2_480-S | - | - |
