Assay Details
| Gene Symbol: Rdh9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153133.2 | NP_694773.1 | 4 | 1926 | 136 | |
| GenBank mRNA | AB081322.1 | - | 4 | 1865 | 136 | |
| AF372838.1 | - | 4 | 1926 | 136 | ||
| AK132181.1 | - | 1997 | 136 | |||
| BC141318.1 | - | 4 | 1905 | 9055930 | 136 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177762 | BC141318.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177762 | AF372838.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177762 | AK132181.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177762 | NM_153133.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177762 | AB081322.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177762 | BC141318.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177762 | AF372838.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177762 | AK132181.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177762 | NM_153133.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177762 | AB081322.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 57286 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 516060 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 353444 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955822 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579745 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883008 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 886048 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254271 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 113555 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 149384 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132948 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100872 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177762 | BC141318.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177762 | AF372838.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177762 | AK132181.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177762 | NM_153133.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177762 | AB081322.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177762 | BC141318.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177762 | AF372838.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177762 | AK132181.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177762 | NM_153133.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177762 | AB081322.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177762 | BC141318.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177762 | AF372838.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177762 | AK132181.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177762 | NM_153133.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177762 | AB081322.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177762 | BC141318.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177762 | AF372838.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177762 | AK132181.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177762 | NM_153133.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P177762 | AB081322.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27538062 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07231370 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02246332 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100872 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07231385 | - | - |
| Whole Mouse Genome G4122A | A_52_P487040 | BC056642.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177762 | BC141318.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177762 | AF372838.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177762 | AK132181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177762 | NM_153133.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177762 | AB081322.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177762 | BC141318.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177762 | AF372838.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177762 | AK132181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177762 | NM_153133.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177762 | AB081322.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113255 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124648 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100872 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07231375 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08635415 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08635429 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08635427 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08635417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27538062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08349660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08635427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27538075 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10367043 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10367044 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427963_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10367041 | - | - |
| Mouse Genome 430 2.0 Array | 1427963_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1427963_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103142_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 103142_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000011333.mm.1 | NM_153133.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011333.mm.1 | NM_153133.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011333.mm.1 | TC1000001573.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011337.mm.1 | NM_153133.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011337.mm.1 | NM_153133.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011337.mm.1 | TC1000001573.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011336.mm.1 | NM_153133.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011336.mm.1 | NM_153133.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011336.mm.1 | TC1000001573.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011338.mm.1 | NM_153133.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011338.mm.1 | NM_153133.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011338.mm.1 | TC1000001573.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011332.mm.1 | NM_153133.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011332.mm.1 | NM_153133.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011332.mm.1 | TC1000001573.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011334.mm.1 | NM_153133.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011334.mm.1 | NM_153133.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011334.mm.1 | TC1000001573.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011340.mm.1 | NM_153133.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011340.mm.1 | NM_153133.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011340.mm.1 | TC1000001573.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011339.mm.1 | NM_153133.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011339.mm.1 | NM_153133.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011339.mm.1 | TC1000001573.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427963_PM_s_at | - | - |
| Mouse Expression Set 430 | 1427963_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427963_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427963_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427963_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427963_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 760316 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2810195 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002810195 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1254902 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254902 | - | - |
| mouse-6 v1.1 (Illumina) | scl38308.4.1_188-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2810195 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38308.4.1_188-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2810195 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1254902 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254902 | - | - |

Rdh9
Genomic Map