Assay Details
| Gene Symbol: Olfr350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146627.1 | NP_666838.1 | 1 | 740 | 139 | |
| GenBank mRNA | BC106790.1 | - | 1 | 740 | 139 | |
| BC106791.1 | - | 1 | 740 | 139 | ||
| BC119406.2 | - | 1 | 740 | 40041609 | 139 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P345261 | NM_146627.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P345261 | BC106790.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P345261 | BC106791.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P345261 | BC119406.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P345261 | NM_146627.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P345261 | BC106790.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P345261 | BC106791.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P345261 | BC119406.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124852 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P345261 | NM_146627.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P345261 | BC106790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P345261 | BC106791.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P345261 | BC119406.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P345261 | NM_146627.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P345261 | BC106790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P345261 | BC106791.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P345261 | BC119406.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P345261 | NM_146627.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P345261 | BC106790.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P345261 | BC106791.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P345261 | BC119406.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P345261 | NM_146627.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P345261 | BC106790.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P345261 | BC106791.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P345261 | BC119406.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140194 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124852 | - | - |
| Whole Mouse Genome G4122A | A_52_P345261 | NM_146627.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P345261 | BC106790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P345261 | BC106791.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P345261 | BC119406.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P345261 | NM_146627.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P345261 | BC106790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P345261 | BC106791.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P345261 | BC119406.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106842 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124852 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01453595 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01453602 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01453584 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01862444 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01857882 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01857881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01601273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02158359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01382588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02112642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01966762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01955786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01966764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01955787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21140238 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10471760 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10471759 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 340715 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3830735 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3830735 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003830735 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2631106 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2631106 | - | - |
| mouse-6 v1.1 (Illumina) | scl21009.1.1_229-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3830735 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21009.1.1_229-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3830735 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2631106 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2631106 | - | - |

Olfr350
Genomic Map