Assay Details
Gene Symbol: Rpl37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026069.3 | NP_080345.1 | 4 | 677 | 125 | |
GenBank mRNA | AK011101.1 | - | 4 | 638 | 125 | |
AK014003.1 | - | 4 | 646 | 125 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982094 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982087 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982093 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982096 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982091 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982088 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982089 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982095 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P06061 | AK011101.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P06061 | AK012544.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P06061 | AK014003.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P06061 | AK019020.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P06061 | AK019294.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211818 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168359 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P06061 | AK011101.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P06061 | AK012544.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P06061 | AK014003.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P06061 | AK019020.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P06061 | AK019294.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206665 | AK011101.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206665 | AK014003.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P206665 | NM_026069.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P150675 | AK011101.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P150675 | AK012544.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P150675 | AK014003.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P150675 | AK019020.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P150675 | AK019294.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982088 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982086 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982099 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982116 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982107 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982093 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982095 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982097 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982094 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982113 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982106 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982076 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982085 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982074 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982069 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982079 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982084 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982100 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982104 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982115 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982105 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982109 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982080 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982091 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982103 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982096 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982081 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29982073 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168359 | - | - |
Whole Mouse Genome G4122A | A_52_P150675 | AK011101.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P150675 | AK012544.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P150675 | AK014003.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P150675 | AK019020.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P150675 | AK019294.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P206665 | AK011101.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P206665 | AK014003.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P206665 | NM_026069.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109613 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168359 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12433450 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12433462 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12433447 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12433460 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12433445 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12433465 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12433478 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982096 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982091 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982095 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982094 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982093 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982087 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982088 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29982089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29982112 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12447576 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982063 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982081 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982117 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982066 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982139 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12433471 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982100 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982088 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982062 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982143 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982086 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982119 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982129 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12135437 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982099 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982102 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982056 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982080 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982057 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982047 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982085 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982048 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982096 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982084 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982097 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11940433 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12447571 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982070 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982109 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982111 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982069 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12377671 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982125 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982061 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982079 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12341324 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982073 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982118 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982124 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982077 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982065 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982137 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12433472 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982095 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982072 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982135 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982068 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982107 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982053 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982060 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982112 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982049 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982110 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12467324 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12341326 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12433443 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982045 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29982103 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10422706 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532924 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532925 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10422702 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10422703 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10422705 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10422704 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1453729_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1434872_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450838_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438291_PM_x_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10422701 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10532923 | - | - |
Mouse Genome 430 2.0 Array | 1450838_x_at | - | - |
Mouse Genome 430 2.0 Array | 1450838_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1453729_a_at | - | - |
Mouse Genome 430 2.0 Array | 1453729_PM_a_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1500000166.mm.1 | NM_026069.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500000166.mm.1 | NM_026069.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500000166.mm.1 | TC1500000019.mm.1 | MTA TC |
Mouse Expression Set 430 | 1453729_a_at | - | - |
Mouse Expression Set 430 | 1453729_PM_a_at | - | - |
Mouse Expression Set 430 | 1434872_x_at | - | - |
Mouse Expression Set 430 | 1450838_PM_x_at | - | - |
Mouse Expression Set 430 | 1450838_x_at | - | - |
Mouse Expression Set 430 | 1438291_PM_x_at | - | - |
Mouse Expression Set 430 | 1434872_PM_x_at | - | - |
Mouse Expression Set 430 | 1438291_x_at | - | - |
Murine Genome U74v2 Set | 92577_PM_f_at | - | - |
Murine Genome U74v2 Set | 92577_f_at | - | - |
Murine Genome U74v2 Set | 168190_PM_at | - | - |
Murine Genome U74v2 Set | 168190_at | - | - |
Murine Genome U74v2 Set | 170891_at | - | - |
Murine Genome U74v2 Set | 170891_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1453729_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1450838_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1434872_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1438291_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1450838_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1434872_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1438291_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1453729_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1453729_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1453729_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 853969 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3170494 | - | - |
MouseWG-6 v2 Expression BeadChip | 3870750 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3870750 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4280494 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3170494 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004280494 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003870750 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003170494 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223052 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2696977 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1236446 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2696977 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1236446 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1223052 | - | - |
mouse-6 v1.1 (Illumina) | scl067281.3_0-S | - | - |
mouse-6 v1.1 (Illumina) | scl48130.4_110-S | - | - |
mouse-6 v1.1 (Illumina) | scl0067281.1_197-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4280494 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3170494 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3870750 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067281.1_197-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl48130.4_110-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl067281.3_0-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3870750 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3170494 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4280494 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1223052 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2696977 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1236446 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2696977 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1236446 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223052 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067281.3_0-S | - | - |
