Assay Details
| Gene Symbol: Atp1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007502.4 | NP_031528.1 | 1 | 117 | ||
| GenBank mRNA | AK032516.1 | - | 1 | 117 | ||
| AK148495.1 | - | 1 | 117 | |||
| AK151700.1 | - | 1 | 117 | |||
| AK151841.1 | - | 1 | 117 | |||
| AK159532.1 | - | 1 | 117 | |||
| AK161282.1 | - | 1 | 117 | |||
| AK168273.1 | - | 1 | 117 | |||
| BC079916.1 | - | 1 | 30665138 | 117 | ||
| U59761.1 | - | 1 | 117 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK032516.1, AK148495.1, AK151700.1, AK151841.1, AK159532.1, AK168273.1, BC079916.1, NM_007502.4, U59761.1 transcripts.
Set Membership: |
|

Atp1b3
Genomic Map