Assay Details
Gene Symbol: Capza3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007605.4 | NP_031631.3 | 1 | 1157 | 129 | |
GenBank mRNA | AK016391.1 | - | 1 | 1126 | 129 | |
BX635805.1 | - | 1 | 129 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302087 | BC049620.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302087 | D87471.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302087 | NM_007605.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302087 | AK016391.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302087 | BC049620.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302087 | D87471.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302087 | NM_007605.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302087 | AK016391.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827472 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150344 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302087 | BC049620.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302087 | D87471.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302087 | NM_007605.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302087 | AK016391.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302087 | BC049620.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302087 | D87471.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302087 | NM_007605.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302087 | AK016391.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302087 | BC049620.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302087 | D87471.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302087 | NM_007605.4 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302087 | AK016391.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302087 | BC049620.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302087 | D87471.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302087 | NM_007605.4 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302087 | AK016391.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892516 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892514 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892524 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892503 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892507 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892505 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892528 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892526 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892519 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892494 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892499 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892496 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892518 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892512 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892509 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892500 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892501 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892520 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892522 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24892502 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150344 | - | - |
Whole Mouse Genome G4122A | A_51_P302087 | BC049620.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302087 | D87471.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302087 | NM_007605.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302087 | AK016391.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302087 | BC049620.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302087 | D87471.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302087 | NM_007605.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302087 | AK016391.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116909 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150344 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05376580 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05376589 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05376576 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05376579 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05376585 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892566 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24892512 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892550 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892508 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892502 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05119150 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05376571 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05090077 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05529566 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05119151 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892521 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892531 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05405884 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892543 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892509 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892501 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05405886 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892535 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892528 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892532 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892518 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892499 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892496 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892553 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892505 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892512 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892540 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05050029 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892538 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05752054 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892525 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24892513 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10542517 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10542516 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418556_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10542515 | - | - |
Mouse Genome 430 2.0 Array | 1418556_at | - | - |
Mouse Genome 430 2.0 Array | 1418556_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12344_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12344_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600013160.mm.1 | TC0600001650.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600013160.mm.1 | NM_007605.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600013160.mm.1 | NM_007605.4 | Refseq |
Mouse Expression Set 430 | 1418556_PM_at | - | - |
Mouse Expression Set 430 | 1418556_at | - | - |
Murine Genome U74v2 Set | 102301_at | - | - |
Murine Genome U74v2 Set | 102301_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418556_at | - | - |
Mouse Genome 430A 2.0 Array | 1418556_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418556_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418556_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 867432 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5670551 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5670551 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005670551 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2618766 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2618766 | - | - |
mouse-6 v1.1 (Illumina) | scl29406.2.26_43-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5670551 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl29406.2.26_43-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5670551 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2618766 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2618766 | - | - |
