Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200020467.mm.1 |
NM_021363.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020467.mm.1 |
NM_021363.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020467.mm.1 |
TC0200002509.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200020465.mm.1 |
NM_021363.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020465.mm.1 |
NM_021363.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020465.mm.1 |
TC0200002509.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200020468.mm.1 |
NM_021363.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020468.mm.1 |
NM_021363.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020468.mm.1 |
TC0200002509.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200020469.mm.1 |
NM_021363.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020469.mm.1 |
NM_021363.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020469.mm.1 |
TC0200002509.mm.1 |
MTA TC |