Assay Details
| Gene Symbol: Cebpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007679.4 | NP_031705.3 | 1 | 1574 | 152 | |
| GenBank mRNA | AK053190.1 | - | 1 | 1574 | 152 | |
| BC139293.1 | - | 1 | 1574 | 8862315 | 152 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
hematopoietic progenitor cell differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
fat cell differentiation
positive regulation of osteoblast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
inner ear development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein homodimerization activity
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
hematopoietic progenitor cell differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
fat cell differentiation
positive regulation of osteoblast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
inner ear development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein homodimerization activity
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567427 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567439 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567437 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567425 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567435 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567436 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567434 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567433 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567438 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567429 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567430 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P444447 | BC139293.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P444447 | X61800.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P444447 | AK156762.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P444447 | AK053190.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P444447 | AK149723.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P444447 | BC139293.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P444447 | X61800.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P444447 | AK156762.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P444447 | AK053190.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P444447 | AK149723.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 417330 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132549 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151533 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P444447 | BC139293.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P444447 | X61800.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P444447 | AK156762.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P444447 | AK053190.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P444447 | AK149723.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P444447 | BC139293.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P444447 | X61800.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P444447 | AK156762.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P444447 | AK053190.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P444447 | AK149723.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P444447 | BC139293.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P444447 | X61800.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P444447 | AK156762.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P444447 | AK053190.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P444447 | AK149723.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P444447 | BC139293.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P444447 | X61800.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P444447 | AK156762.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P444447 | AK053190.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P444447 | AK149723.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30567446 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132549 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151533 | - | - |
| Whole Mouse Genome G4122A | A_51_P444447 | BC139293.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444447 | X61800.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444447 | AK156762.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444447 | AK053190.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444447 | AK149723.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444447 | BC139293.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444447 | X61800.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444447 | AK156762.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444447 | AK053190.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444447 | AK149723.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114637 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151533 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132549 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567437 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567436 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567435 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567439 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567433 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567427 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567425 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567429 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567430 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567434 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567438 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30567432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30567423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12620361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30567415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12620372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30567442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12786122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30567424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30567423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30567431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12620365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30567440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30567419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30567449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30567421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12924097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30567418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12786107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30567413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30567444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30567438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30567425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30567447 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10433886 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456605_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423233_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10433885 | - | - |
| Mouse Genome 430 2.0 Array | 1423233_at | - | - |
| Mouse Genome 430 2.0 Array | 1456605_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456605_at | - | - |
| Mouse Genome 430 2.0 Array | 1423233_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12609_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12609_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600001487.mm.1 | NM_007679.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001487.mm.1 | NM_007679.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001487.mm.1 | TC1600000192.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600001488.mm.1 | NM_007679.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001488.mm.1 | NM_007679.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001488.mm.1 | TC1600000192.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600001489.mm.1 | NM_007679.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001489.mm.1 | NM_007679.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001489.mm.1 | TC1600000192.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456605_PM_at | - | - |
| Mouse Expression Set 430 | 1423233_at | - | - |
| Mouse Expression Set 430 | 1456605_at | - | - |
| Mouse Expression Set 430 | 1423233_PM_at | - | - |
| Murine Genome U74v2 Set | 160894_PM_at | - | - |
| Murine Genome U74v2 Set | 160894_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456605_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423233_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456605_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423233_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456605_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456605_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423233_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423233_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5700017 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5700017 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005700017 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2707363 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2588570 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2588570 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2707363 | - | - |
| mouse-6 v1.1 (Illumina) | scl49391.2.1_191-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5700017 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49391.2.1_191-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5700017 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2588570 | - | - |

Cebpd
Genomic Map