Assay Details
| Gene Symbol: Ndufb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025843.3 | NP_080119.1 | 3 | 479 | 136 | |
| GenBank mRNA | AK002678.1 | - | 3 | 477 | 136 | |
| AK003300.1 | - | 3 | 480 | 136 | ||
| BC034787.1 | - | 3 | 458 | 4913656 | 136 | |
| BC056984.1 | - | 2 | 4320 | 136 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26015452 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26015447 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26015450 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26015449 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P519276 | NM_025843.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P519276 | AK002678.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P519276 | AK003300.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P519276 | BC034787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P519276 | BC056984.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P519276 | NM_025843.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P519276 | AK002678.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P519276 | AK003300.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P519276 | BC034787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P519276 | BC056984.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 370293 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 934005 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 95079 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 485698 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 475192 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 328772 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 612877 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137440 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84379 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 173361 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 152431 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168816 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P519276 | NM_025843.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P519276 | AK002678.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P519276 | AK003300.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P519276 | BC034787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P519276 | BC056984.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P519276 | NM_025843.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P519276 | AK002678.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P519276 | AK003300.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P519276 | BC034787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P519276 | BC056984.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519276 | NM_025843.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519276 | AK002678.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519276 | AK003300.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519276 | BC034787.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519276 | BC056984.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519276 | NM_025843.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519276 | AK002678.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519276 | AK003300.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519276 | BC034787.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519276 | BC056984.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26015441 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168816 | - | - |
| Whole Mouse Genome G4122A | A_51_P519276 | NM_025843.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519276 | AK002678.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519276 | AK003300.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519276 | BC034787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519276 | BC056984.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519276 | NM_025843.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519276 | AK002678.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519276 | AK003300.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519276 | BC034787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519276 | BC056984.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113568 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168816 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06820167 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07259812 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07259814 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07416067 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06745305 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26015449 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26015452 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26015450 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26015447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26015461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07259833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07263624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07416067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06710189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07130022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07259814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07259818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07356110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06918962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26015472 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10573195 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573196 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573197 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416417_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448331_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10573194 | - | - |
| Mouse Genome 430 2.0 Array | 1448331_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448331_at | - | - |
| Mouse Genome 430 2.0 Array | 1416417_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416417_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66916_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66916_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800006839.mm.1 | NM_025843.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006839.mm.1 | NM_025843.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006839.mm.1 | TC0800000926.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800006841.mm.1 | NM_025843.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006841.mm.1 | NM_025843.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006841.mm.1 | TC0800000926.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800006842.mm.1 | NM_025843.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006842.mm.1 | NM_025843.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006842.mm.1 | TC0800000926.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448331_PM_at | - | - |
| Mouse Expression Set 430 | 1448331_at | - | - |
| Mouse Expression Set 430 | 1416417_PM_a_at | - | - |
| Mouse Expression Set 430 | 1416417_a_at | - | - |
| Murine Genome U74v2 Set | 95698_at | - | - |
| Murine Genome U74v2 Set | 162323_at | - | - |
| Murine Genome U74v2 Set | 95698_PM_at | - | - |
| Murine Genome U74v2 Set | 162323_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448331_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448331_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416417_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416417_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448331_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416417_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448331_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416417_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 739525 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1850026 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1850026 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001850026 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2717460 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2717460 | - | - |
| mouse-6 v1.1 (Illumina) | scl066916.1_19-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1850026 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066916.1_19-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1850026 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2717460 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2717460 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066916.1_19-S | - | - |

Ndufb7
Genomic Map