Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100009983.mm.1 |
NM_007695.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009983.mm.1 |
NM_007695.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009983.mm.1 |
TC0100001222.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100009989.mm.1 |
NM_007695.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009989.mm.1 |
NM_007695.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009989.mm.1 |
TC0100001222.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100009988.mm.1 |
NM_007695.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009988.mm.1 |
NM_007695.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009988.mm.1 |
TC0100001222.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100009992.mm.1 |
NM_007695.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009992.mm.1 |
NM_007695.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100009992.mm.1 |
TC0100001222.mm.1 |
MTA TC |