Assay Details
Gene Symbol: Cr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007758.2 | NP_031784.1 | 3 - 4 | 747 | 77 | |
GenBank mRNA | AK003072.2 | - | 3 - 4 | 747 | 77 | |
BC109344.1 | - | 3 - 4 | 680 | 40054528 | 77 | |
M35684.1 | - | 2 - 3 | 615 | 77 | ||
M61132.1 | - | 2 - 3 | 582 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
