Assay Details
Gene Symbol: Hint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008248.2 | NP_032274.1 | 1 - 2 | 167 | 84 | |
GenBank mRNA | AK002965.2 | - | 1 - 2 | 167 | 84 | |
AK012433.1 | - | 1 - 2 | 173 | 84 | ||
BC070415.1 | - | 1 - 2 | 212 | 5714321 | 84 | |
BC080296.1 | - | 1 - 2 | 151 | 6814185 | 84 | |
U60001.1 | - | 1 - 2 | 137 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
purine ribonucleotide catabolic process
intrinsic apoptotic signaling pathway by p53 class mediator
nucleotide binding
catalytic activity
GPI-anchor transamidase activity
hydrolase activity
coenzyme F390-A hydrolase activity
coenzyme F390-G hydrolase activity
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
purine ribonucleotide catabolic process
intrinsic apoptotic signaling pathway by p53 class mediator
nucleotide binding
catalytic activity
GPI-anchor transamidase activity
hydrolase activity
coenzyme F390-A hydrolase activity
coenzyme F390-G hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27812450 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27812446 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27812448 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27812449 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27812447 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192860 | BC070415.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192860 | BC080296.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192860 | U60001.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192860 | NM_008248.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192860 | AK012433.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192860 | BC070415.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192860 | BC080296.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192860 | U60001.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192860 | NM_008248.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192860 | AK012433.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96709 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508240 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76244 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144487 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192860 | BC070415.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192860 | BC080296.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192860 | U60001.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192860 | NM_008248.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192860 | AK012433.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192860 | BC070415.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192860 | BC080296.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192860 | U60001.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192860 | NM_008248.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192860 | AK012433.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P192860 | BC070415.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P192860 | BC080296.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P192860 | U60001.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P192860 | NM_008248.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P192860 | AK012433.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P192860 | BC070415.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P192860 | BC080296.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P192860 | U60001.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P192860 | NM_008248.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P192860 | AK012433.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27812461 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27812455 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27812458 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27812459 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27812447 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27812450 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27812453 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27812457 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27812445 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144487 | - | - |
Whole Mouse Genome G4122A | A_51_P192860 | BC070415.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P192860 | BC080296.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P192860 | U60001.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P192860 | NM_008248.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P192860 | AK012433.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P192860 | BC070415.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P192860 | BC080296.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P192860 | U60001.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P192860 | NM_008248.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P192860 | AK012433.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119003 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144487 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27812447 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27812450 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27812449 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27812446 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27812448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27812473 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812466 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812469 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812473 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812500 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812457 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09660851 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812507 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812488 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09712349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812462 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09529544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09395222 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812453 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812479 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812437 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09018362 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09407863 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812482 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812450 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812434 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812504 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812442 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09185089 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812471 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812464 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812451 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812438 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812445 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812474 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812481 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812486 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09407861 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812484 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812443 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09248860 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812499 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812459 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812436 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812435 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812461 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812440 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27812467 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10376200 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10376199 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10376197 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10376198 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1424017_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1424018_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10376196 | - | - |
Mouse Genome 430 2.0 Array | 1457286_at | - | - |
Mouse Genome 430 2.0 Array | 1424018_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1424017_a_at | - | - |
Mouse Genome 430 2.0 Array | 1424018_at | - | - |
Mouse Genome 430 2.0 Array | 1424017_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1457286_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15254_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15254_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100005141.mm.1 | TC1100000637.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100005144.mm.1 | NM_008248.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100005144.mm.1 | NM_008248.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100005144.mm.1 | TC1100000637.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100005145.mm.1 | NM_008248.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100005145.mm.1 | NM_008248.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100005145.mm.1 | TC1100000637.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100005143.mm.1 | NM_008248.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100005143.mm.1 | NM_008248.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100005143.mm.1 | TC1100000637.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100005148.mm.1 | NM_008248.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100005148.mm.1 | NM_008248.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100005148.mm.1 | TC1100000637.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100005147.mm.1 | NM_008248.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100005147.mm.1 | NM_008248.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100005147.mm.1 | TC1100000637.mm.1 | MTA TC |
Mouse Expression Set 430 | 1424017_a_at | - | - |
Mouse Expression Set 430 | 1424017_PM_a_at | - | - |
Mouse Expression Set 430 | 1424018_at | - | - |
Mouse Expression Set 430 | 1424018_PM_at | - | - |
Murine Genome U74v2 Set | 99581_at | - | - |
Murine Genome U74v2 Set | 99581_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1424017_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1424017_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1424018_at | - | - |
Mouse Genome 430A 2.0 Array | 1424018_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424017_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424018_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424017_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424018_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 431274 | - | - |
ABI Mouse Genome Survey Microarray | 893708 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2640128 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2640128 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002640128 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2723631 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2723631 | - | - |
mouse-6 v1.1 (Illumina) | scl41548.2.100_36-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2640128 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl41548.2.100_36-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2640128 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2723631 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2723631 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl41548.2.100_36-S | - | - |
