Assay Details
| Gene Symbol: Apbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201413.1 | NP_001188342.1 | 2 - 3 | 412 | 63 | |
| NM_001201415.1 | NP_001188344.1 | 2 - 3 | 412 | 63 | ||
| NM_009686.2 | NP_033816.1 | 2 - 3 | 412 | 63 | ||
| GenBank mRNA | AK004792.1 | - | 2 - 3 | 266 | 63 | |
| AK032770.1 | - | 2 - 3 | 278 | 63 | ||
| AK167649.1 | - | 2 - 3 | 277 | 63 | ||
| AK169495.1 | - | 2 - 3 | 265 | 63 | ||
| AK169959.1 | - | 2 - 3 | 353 | 63 | ||
| AK183807.1 | - | 1 - 2 | 174 | 63 | ||
| AK196430.1 | - | 1 - 2 | 77 | 63 | ||
| BC076587.1 | - | 2 - 3 | 258 | 30550087 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK004792.1, AK032770.1, AK167649.1, AK169495.1, AK169959.1, BC076587.1, NM_001201413.1, NM_001201415.1, NM_009686.2 transcripts.
Set Membership: |
|

Apbb2
Genomic Map