Assay Details
Gene Symbol: Cryz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009968.3 | NP_034098.1 | 4 - 5 | 1016 | 63 | |
GenBank mRNA | AK098082.1 | - | 4 - 5 | 500 | 63 | |
AK133820.1 | - | 4 - 5 | 1017 | 63 | ||
AK134281.1 | - | 4 - 5 | 501 | 63 | ||
BC003800.1 | - | 4 - 5 | 508 | 3492859 | 63 | |
BC043076.1 | - | 4 - 5 | 477 | 6404996 | 63 | |
D78646.1 | - | 1 - 2 | 43 | 63 | ||
S70056.1 | - | 4 - 5 | 528 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
