Assay Details
| Gene Symbol: Cst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009978.2 | NP_034108.1 | 3 - 4 | 535 | 116 | |
| GenBank mRNA | AF090691.1 | - | 3 - 4 | 524 | 116 | |
| AK018943.1 | - | 3 - 4 | 536 | 116 | ||
| AK054409.1 | - | 3 - 4 | 518 | 116 | ||
| AK161808.1 | - | 3 - 4 | 528 | 116 | ||
| BC049753.1 | - | 3 - 4 | 540 | 6773192 | 116 | |
| S49926.1 | - | 3 - 4 | 336 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038106 | AK161808.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038106 | NM_009978.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038106 | AF090691.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038106 | AK018943.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 113914 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569485 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 273136 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 504280 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11144 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128352 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038106 | AK161808.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038106 | NM_009978.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038106 | AF090691.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038106 | AK018943.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21732242 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128352 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04734476 | - | - |
| Whole Mouse Genome G4122A | A_51_P307840 | AK161808.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307840 | BC049753.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307840 | S49926.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307840 | NM_009978.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307840 | AF090691.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134184 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128352 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01366153 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01366143 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02124761 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01366155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21732323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01191153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02094155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01281479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01382132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01701096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01382121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02124761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01191150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01366156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02197461 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01366158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01825537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02094159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01620420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732280 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01281487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01382129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01783831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21732255 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10476916 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476917 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476918 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424479_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10476915 | - | - |
| Mouse Genome 430 2.0 Array | 1424479_at | - | - |
| Mouse Genome 430 2.0 Array | 1424479_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13012_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13012_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200017420.mm.1 | NM_009978.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200017420.mm.1 | NM_009978.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200017420.mm.1 | TC0200002201.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424479_at | - | - |
| Mouse Expression Set 430 | 1424479_PM_at | - | - |
| Murine Genome U74v2 Set | 103245_at | - | - |
| Murine Genome U74v2 Set | 103245_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424479_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424479_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424479_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424479_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 908850 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1660605 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1660605 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001660605 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2707745 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2707745 | - | - |
| mouse-6 v1.1 (Illumina) | scl20154.4.1_28-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1660605 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20154.4.1_28-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1660605 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2707745 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2707745 | - | - |

Cst8
Genomic Map