Assay Details
| Gene Symbol: Asap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276461.1 | NP_001263390.1 | 8 - 9 | 884 | 111 | |
| NM_001276462.1 | NP_001263391.1 | 6 - 7 | 793 | 111 | ||
| NM_001276463.1 | NP_001263392.1 | 6 - 7 | 793 | 111 | ||
| NM_001276467.1 | NP_001263396.1 | 8 - 9 | 884 | 111 | ||
| NM_010026.3 | NP_034156.2 | 8 - 9 | 884 | 111 | ||
| GenBank mRNA | AF075461.1 | - | 8 - 9 | 703 | 111 | |
| AF075462.1 | - | 8 - 9 | 703 | 111 | ||
| AK122477.1 | - | 6 - 7 | 488 | 111 | ||
| AK146545.1 | - | 8 - 9 | 722 | 111 | ||
| AK147689.1 | - | 6 - 7 | 794 | 111 | ||
| BC048818.1 | - | 6 - 7 | 759 | 111 | ||
| BC094581.1 | - | 8 - 9 | 884 | 30549241 | 111 | |
| CD355387.1 | - | 4 - 5 | 381 | 111 | ||
| CN455356.1 | - | 1 - 2 | 108 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Asap1
Genomic Map