Assay Details
Gene Symbol: Des | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010043.2 | NP_034173.1 | 2 - 3 | 757 | 91 | |
GenBank mRNA | AK131922.1 | - | 2 - 3 | 717 | 91 | |
AK132391.1 | - | 2 - 3 | 721 | 91 | ||
BC031760.1 | - | 2 - 3 | 705 | 4219280 | 91 | |
L22550.1 | - | 2 - 3 | 714 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
