Assay Details
| Gene Symbol: Gngt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010314.2 | NP_034444.1 | 1 - 2 | 74 | 110 | |
| GenBank mRNA | AK020863.1 | - | 1 - 2 | 76 | 110 | |
| AK020903.1 | - | 1 - 2 | 75 | 110 | ||
| BC025929.1 | - | 1 - 2 | 269 | 4511672 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK020863.1, AK020903.1, BC025929.1, NM_010314.2 transcripts.
Set Membership: |
|

Gngt1
Genomic Map