Assay Details
Gene Symbol: Lck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162432.1 | NP_001155904.1 | 3 - 4 | 274 | 71 | |
NM_001162433.1 | NP_001155905.1 | 3 - 4 | 316 | 71 | ||
NM_010693.3 | NP_034823.1 | 2 - 3 | 371 | 71 | ||
GenBank mRNA | AK088001.1 | - | 2 - 3 | 372 | 71 | |
AK134013.1 | - | 2 - 3 | 361 | 71 | ||
BC011474.1 | - | 3 - 4 | 281 | 4191610 | 71 | |
BQ563042.1 | - | 3 - 4 | 265 | 71 | ||
M12056.1 | - | 2 - 3 | 347 | 71 | ||
X03533.1 | - | 2 - 3 | 377 | 71 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
