Assay Details
Gene Symbol: Ppp5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011155.2 | NP_035285.2 | 2 - 3 | 461 | 78 | |
GenBank mRNA | AF018262.1 | - | 2 - 3 | 432 | 78 | |
AK180337.1 | - | 2 - 3 | 279 | 78 | ||
BC003744.1 | - | 2 - 3 | 417 | 3590322 | 78 | |
DV653678.1 | - | 2 - 3 | 398 | 78 | ||
U12204.1 | - | 2 - 3 | 329 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
