Assay Details
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011188.3 | NP_035318.1 | 1 - 2 | 229 | 149 | |
GenBank mRNA | AK076874.1 | - | 1 - 2 | 231 | 149 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK076874.1, NM_011188.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
