Assay Details
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011188.3 | NP_035318.1 | 3 - 4 | 505 | 104 | |
GenBank mRNA | AK005083.1 | - | 2 - 3 | 128 | 104 | |
AK076874.1 | - | 3 - 4 | 507 | 104 | ||
AK146919.1 | - | 2 - 3 | 146 | 104 | ||
AK153413.1 | - | 2 - 3 | 149 | 104 | ||
AK168548.1 | - | 2 - 3 | 146 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
