Assay Details
| Gene Symbol: Pstpip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011193.2 | NP_035323.2 | 4 - 5 | 675 | 72 | |
| GenBank mRNA | AK037936.1 | - | 4 - 5 | 677 | 72 | |
| AK088150.1 | - | 4 - 5 | 672 | 72 | ||
| BC096761.1 | - | 4 - 5 | 455 | 30040955 | 72 | |
| U87814.1 | - | 4 - 5 | 515 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pstpip1
Genomic Map