Assay Details
| Gene Symbol: Ik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011879.2 | NP_036009.1 | 11 - 12 | 1064 | 105 | |
| GenBank mRNA | AJ006130.1 | - | 11 - 12 | 947 | 105 | |
| AK029644.1 | - | 11 - 12 | 1018 | 105 | ||
| AK050625.1 | - | 11 - 12 | 1022 | 105 | ||
| AK082356.1 | - | 11 - 12 | 996 | 105 | ||
| AK087976.1 | - | 11 - 12 | 1010 | 105 | ||
| BC006706.1 | - | 9 - 10 | 1422 | 105 | ||
| BC014739.1 | - | 11 - 12 | 995 | 4920184 | 105 | |
| BC026569.1 | - | 11 - 12 | 995 | 4982612 | 105 | |
| BC029671.1 | - | 11 - 12 | 1007 | 5364953 | 105 | |
| BC029694.1 | - | 11 - 12 | 1004 | 5369477 | 105 | |
| BC033356.1 | - | 11 - 12 | 991 | 5366230 | 105 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ik
Genomic Map