Assay Details
| Gene Symbol: Ik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011879.2 | NP_036009.1 | 17 - 18 | 1619 | 67 | |
| GenBank mRNA | AJ006130.1 | - | 17 - 18 | 1502 | 67 | |
| AK087976.1 | - | 17 - 18 | 1565 | 67 | ||
| BC006706.1 | - | 15 - 16 | 1977 | 67 | ||
| BC014739.1 | - | 17 - 18 | 1550 | 4920184 | 67 | |
| BC026569.1 | - | 17 - 18 | 1550 | 4982612 | 67 | |
| BC029148.1 | - | 15 - 16 | 2513 | 67 | ||
| BC029671.1 | - | 17 - 18 | 1562 | 5364953 | 67 | |
| BC029694.1 | - | 17 - 18 | 1559 | 5369477 | 67 | |
| BC033356.1 | - | 17 - 18 | 1546 | 5366230 | 67 | |
| BM211698.2 | - | 3 - 4 | 67 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ik
Genomic Map