Assay Details
Gene Symbol: Map3k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011945.2 | NP_036075.2 | 17 - 18 | 4077 | 91 | |
GenBank mRNA | AF117340.1 | - | 17 - 18 | 4077 | 91 | |
L13103.1 | - | 7 - 8 | 2086 | 91 | ||
U23470.1 | - | 4 - 5 | 1678 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
