Assay Details
| Gene Symbol: Map3k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011947.3 | NP_036077.1 | 14 - 15 | 1506 | 65 | |
| GenBank mRNA | AK137127.1 | - | 14 - 15 | 1792 | 65 | |
| AK220450.1 | - | 13 - 14 | 2396 | 65 | ||
| BC023781.1 | - | 14 - 15 | 1510 | 5346982 | 65 | |
| U43187.1 | - | 14 - 15 | 1809 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Map3k3
Genomic Map