Assay Details
| Gene Symbol: Angptl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013913.3 | NP_038941.1 | 5 - 6 | 985 | 117 | |
| GenBank mRNA | AF162224.1 | - | 5 - 6 | 953 | 117 | |
| AK149450.1 | - | 5 - 6 | 986 | 117 | ||
| AK149555.1 | - | 5 - 6 | 986 | 117 | ||
| BC019491.1 | - | 5 - 6 | 973 | 4211688 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Angptl3
Genomic Map