Assay Details
| Gene Symbol: Rbpms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286168.1 | NP_001273097.1 | 6 - 7 | 787 | 80 | |
| NM_019733.3 | NP_062707.1 | 7 - 8 | 921 | 80 | ||
| GenBank mRNA | AF148511.1 | - | 7 - 8 | 870 | 80 | |
| AK079238.1 | - | 6 - 7 | 731 | 80 | ||
| BC030397.1 | - | 7 - 8 | 882 | 4225078 | 80 | |
| BY708385.1 | - | 4 - 5 | 415 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbpms
Genomic Map