Assay Details
| Gene Symbol: Slc12a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020333.2 | NP_065066.2 | 4 - 5 | 842 | 76 | |
| GenBank mRNA | AF332063.1 | - | 4 - 5 | 512 | 76 | |
| AF332064.1 | - | 4 - 5 | 512 | 76 | ||
| AK122460.1 | - | 4 - 5 | 576 | 76 | ||
| AK147262.1 | - | 4 - 5 | 844 | 76 | ||
| AK165769.1 | - | 4 - 5 | 590 | 76 | ||
| BC054808.1 | - | 4 - 5 | 508 | 6838880 | 76 | |
| KJ535320.1 | - | 4 - 5 | 533 | 76 | ||
| KJ535321.1 | - | 4 - 5 | 533 | 76 | ||
| KJ535322.1 | - | 4 - 5 | 689 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc12a5
Genomic Map