Assay Details
| Gene Symbol: Ssbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023672.2 | NP_076161.2 | 4 - 5 | 666 | 119 | |
| NM_198438.1 | NP_940840.1 | 4 - 5 | 666 | 119 | ||
| GenBank mRNA | AF170906.1 | - | 4 - 5 | 396 | 119 | |
| AF500117.1 | - | 4 - 5 | 396 | 119 | ||
| AK011853.1 | - | 4 - 5 | 636 | 119 | ||
| AK211107.1 | - | 2 - 3 | 116 | 119 | ||
| AY167987.1 | - | 4 - 5 | 279 | 119 | ||
| BC003430.1 | - | 4 - 5 | 420 | 2646149 | 119 | |
| BC035475.1 | - | 4 - 5 | 609 | 119 | ||
More Information
Set Membership: |
|

Ssbp3
Genomic Map