Assay Details
| Gene Symbol: Ssbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023672.2 | NP_076161.2 | 5 - 6 | 755 | 51 | |
| GenBank mRNA | AK011509.1 | - | 5 - 6 | 373 | 51 | |
| AK011853.1 | - | 5 - 6 | 725 | 51 | ||
| AK133812.1 | - | 2 - 3 | 207 | 51 | ||
More Information
Set Membership: |
|

Ssbp3
Genomic Map