Assay Details
| Gene Symbol: Ssbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023672.2 | NP_076161.2 | 6 - 7 | 832 | 62 | |
| GenBank mRNA | AK011509.1 | - | 6 - 7 | 450 | 62 | |
| AK011853.1 | - | 6 - 7 | 802 | 62 | ||
| AK017711.1 | - | 6 - 7 | 835 | 62 | ||
| AK133812.1 | - | 3 - 4 | 284 | 62 | ||
More Information
Set Membership: |
|

Ssbp3
Genomic Map