Assay Details
| Gene Symbol: Ssbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023672.2 | NP_076161.2 | 8 - 9 | 958 | 66 | |
| NM_198438.1 | NP_940840.1 | 7 - 8 | 877 | 66 | ||
| GenBank mRNA | AF170906.1 | - | 7 - 8 | 607 | 66 | |
| AF500117.1 | - | 7 - 8 | 607 | 66 | ||
| AK011509.1 | - | 8 - 9 | 576 | 66 | ||
| AK011853.1 | - | 8 - 9 | 928 | 66 | ||
| AK017711.1 | - | 8 - 9 | 961 | 66 | ||
| AK041388.1 | - | 4 - 5 | 474 | 66 | ||
| AK133812.1 | - | 5 - 6 | 411 | 66 | ||
| AK153733.1 | - | 4 - 5 | 318 | 66 | ||
| AY167987.1 | - | 7 - 8 | 490 | 66 | ||
| BC003430.1 | - | 7 - 8 | 631 | 2646149 | 66 | |
| BC028324.1 | - | 2 - 3 | 99 | 66 | ||
| BC035475.1 | - | 7 - 8 | 820 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC028324.1 transcripts.
Set Membership: |
|

Ssbp3
Genomic Map