Assay Details
Gene Symbol: Kin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025280.2 | NP_079556.1 | 3 - 4 | 272 | 137 | |
GenBank mRNA | AK009429.1 | - | 3 - 4 | 304 | 137 | |
AK132548.1 | - | 3 - 4 | 702 | 137 | ||
AK167740.1 | - | 3 - 4 | 276 | 137 | ||
BC028860.1 | - | 3 - 4 | 272 | 3673830 | 137 | |
BC058169.1 | - | 3 - 4 | 301 | 137 | ||
X58472.1 | - | 3 - 4 | 277 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
