Assay Details
Gene Symbol: Mrpl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177658.1 | NP_001171129.1 | 2 - 3 | 310 | 93 | |
NM_025300.4 | NP_079576.1 | 2 - 3 | 310 | 93 | ||
GenBank mRNA | AK003367.1 | - | 2 - 3 | 294 | 93 | |
AK010474.1 | - | 2 - 3 | 270 | 93 | ||
AK010604.1 | - | 2 - 3 | 303 | 93 | ||
AK017820.1 | - | 2 - 3 | 289 | 93 | ||
AK017824.1 | - | 2 - 3 | 273 | 93 | ||
AK019175.1 | - | 2 - 3 | 283 | 93 | ||
AK019244.1 | - | 2 - 3 | 270 | 93 | ||
AK019249.1 | - | 2 - 3 | 270 | 93 | ||
AK150880.1 | - | 2 - 3 | 311 | 93 | ||
AK160660.1 | - | 2 - 3 | 268 | 93 | ||
BC021304.1 | - | 2 - 3 | 306 | 3498041 | 93 | |
BC027233.1 | - | 2 - 3 | 295 | 3498065 | 93 | |
BC068230.1 | - | 2 - 3 | 293 | 93 | ||
BY083200.1 | - | 2 - 3 | 325 | 93 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
