Assay Details
| Gene Symbol: Sh3d21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162533.1 | NP_001156005.1 | 14 - 15 | 1987 | 61 | |
| NM_025856.3 | NP_080132.1 | 10 - 11 | 1651 | 61 | ||
| GenBank mRNA | AK006490.1 | - | 1 - 2 | 772 | 61 | |
| AK076962.1 | - | 10 - 11 | 1616 | 61 | ||
| AK155132.1 | - | 14 - 15 | 1908 | 61 | ||
| BC019568.1 | - | 14 - 15 | 1904 | 61 | ||
| BC025637.1 | - | 8 - 9 | 1648 | 61 | ||
| BC039777.1 | - | 10 - 11 | 1841 | 61 | ||
| BC053421.1 | - | 10 - 11 | 1612 | 6706860 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3d21
Genomic Map