Assay Details
| Gene Symbol: Sh3d21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162533.1 | NP_001156005.1 | 10 - 11 | 882 | 105 | |
| NM_025856.3 | NP_080132.1 | 6 - 7 | 546 | 105 | ||
| GenBank mRNA | AK011430.1 | - | 6 - 7 | 105 | ||
| AK076962.1 | - | 6 - 7 | 511 | 105 | ||
| AK155132.1 | - | 10 - 11 | 803 | 105 | ||
| BC019568.1 | - | 10 - 11 | 799 | 105 | ||
| BC025637.1 | - | 5 - 6 | 456 | 105 | ||
| BC039777.1 | - | 6 - 7 | 736 | 105 | ||
| BC053421.1 | - | 6 - 7 | 507 | 6706860 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011430.1, AK076962.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23278547 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23278550 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23278548 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23278549 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23278552 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983368 | BF452472.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983368 | BC019568.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983368 | BC025637.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983368 | BC039777.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983368 | AK076962.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191948 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824855 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 732752 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713896 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 756405 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 314749 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29535 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147784 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126950 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114982 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983368 | BF452472.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983368 | BC019568.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983368 | BC025637.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983368 | BC039777.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983368 | AK076962.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278593 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23278582 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114982 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05391335 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126950 | - | - |
| Whole Mouse Genome G4122A | A_52_P456192 | BC053421.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P456192 | BC019568.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P456192 | BC025637.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P456192 | BC039777.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P456192 | AK011430.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139109 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114982 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126950 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03443993 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03443987 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03443974 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03443982 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03443976 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03443999 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03443997 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03443979 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03443975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23278547 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23278549 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23278552 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23278550 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23278548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23278520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03378749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03443993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278547 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03935169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278558 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278522 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03378748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278510 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278524 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03935208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03443999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278544 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03443974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03660966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03935190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03444856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23278532 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10516282 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516287 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516279 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516292 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516284 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516278 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516290 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516291 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516288 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516289 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516285 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516294 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516295 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516280 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516286 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516281 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516283 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428298_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10516277 | - | - |
| Mouse Genome 430 2.0 Array | 1428298_at | - | - |
| Mouse Genome 430 2.0 Array | 1428298_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66938_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66938_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400028637.mm.1 | TC0400003529.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400028637.mm.1 | NM_001162533.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400028637.mm.1 | NM_001162533.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400028637.mm.1 | NM_025856.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400028637.mm.1 | NM_025856.3 | Refseq |
| Mouse Expression Set 430 | 1428298_PM_at | - | - |
| Mouse Expression Set 430 | 1428298_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428298_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428298_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 912538 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100870114 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100870114 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5720170 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005720170 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250435 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2546026 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250435 | - | - |
| mouse-6 v1.1 (Illumina) | scl23824.14.1_21-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5720170 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100870114 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23824.14.1_21-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5720170 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100870114 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1250435 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2546026 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250435 | - | - |

Sh3d21
Genomic Map